missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NOD1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11385993
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11385993 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11385993 Supplier Invitrogen™ Supplier No. PA518027

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

NOD1 is a member of the NOD (nucleotide-binding oligomerization domain) family, a group of proteins that are involved in innate immune defense. NOD1 contains an N-terminal caspase recruitment domain (CARD), a centrally located nucleotide-binding domain (NBD), and ten tandem leucine-rich repeats (LRRs) in its C-terminus. The CARD is involved in apoptotic signaling, and NOD1 activates caspase-9 and NF-kappa-B. LRRs participate in protein-protein interactions, and mutations in the NBD may affect the process of oligomerization and subsequent function of the LRR domain. This protein is an intracellular pattern-recognition receptor (PRR) that initiates inflammation in response to a subset of bacteria through the detection of bacterial diaminopimelic acid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOD1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene NOD1
Gene Accession No. Q9Y239
Gene Alias C230079P11; Card4; caspase recruitment domain 4; caspase recruitment domain family, member 4; caspase recruitment domain-containing protein 4; CLR7.1; F830007N14Rik; NLR family, CARD domain containing 1; Nlrc1; Nod1; nucleotide binding oligomerization domain containing 1; nucleotide-binding oligomerization domain containing 1; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 1; nucleotide-binding oligomerization domain-containing protein 1; RGD1562269
Gene Symbols NOD1
Host Species Goat
Immunogen Synthetic peptide sequence (EAKVYEDEKRIICF) corresponding to the C-terminus amino acids of NOD1 (aa 940-953).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10392
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.