missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMUR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA132767
This item is not returnable.
View return policy
Description
PA1-32767 detects NMUR2 from human samples.
This gene encodes a protein from the G-protein coupled receptor 1 family. This protein is a receptor for neuromedin U, which is a neuropeptide that is widely distributed in the gut and central nervous system. This receptor plays an important role in the regulation of food intake and body weight.
Specifications
| NMUR2 | |
| Polyclonal | |
| Unconjugated | |
| Nmur2 | |
| FM4; FM-4; G-protein coupled receptor FM-4; G-protein coupled receptor TGR-1; G-protein-coupled receptor FM-4; growth hormone secretagogue receptor family, member 4; neuromedin s receptor 2 protein; neuromedin U receptor 2; neuromedin-U receptor 2; NMSR2; NMU2R; NMUR2; NMU-R2; Tgr1; TGR-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100217382, 56923 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9GZQ4 | |
| Nmur2 | |
| Synthetic 19 amino acid peptide from 3rd cytoplasmic domain of human NMUR2 | |
| 25 μg | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction