Learn More
N-myc (and STAT) interactor, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human NMI protein.
Brand: Abnova H00009111-D01P.100ug
Description
NMYC interactor (NMI) encodes a protein that interacts with NMYC and CMYC (two members of the oncogene Myc family), and other transcription factors containing a Zip, HLH, or HLH-Zip motif. The NMI protein also interacts with all STATs except STAT2 and augments STAT-mediated transcription in response to cytokines IL2 and IFN-gamma. The NMI mRNA has low expression levels in all human fetal and adult tissues tested except brain and has high expression in cancer cell line-myeloid leukemias. [provided by RefSeq
Sequence: MEADKDDTQQILKEHSPDEFIKDEQNKGLIDEITKKNIQLRKEIQKLETELQEATKEFQIKEDIPETKMKFLSVETPENDSQLSNISCSFQVSSKVPYEIQKGQALITFEKEEVAQNVVSMSKHHVQIKDVNLEVTAKPVPLNSGVRFQVYVEVSKMKINVTEIPDTLREDQMRDKLELSFSKSRNGGGEVDRVDYDRQSGSAVITFVEIGVADKILKKKEYPLYINQTCHRVTVSPYTEIHLKKYQIFSGTSKRTVLLTGMEGIQMDEEIVEDLINIHFQRAKNGGGEVDVVKCSLGQPHIAYFEESpecifications
| N-myc (and STAT) interactor | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human NMI protein. | |
| NMI | |
| NMI | |
| NMI (AAH21987.1, 1 a.a. ∼ 307 a.a) full-length human protein. | |
| 100 μg | |
| Signal Transduction | |
| 9111 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC021987.2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.