Learn More
N-myc (and STAT) interactor, Mouse, Clone: 9B8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant NMI.
Brand: Abnova H00009111-M03.100ug
Description
NMYC interactor (NMI) encodes a protein that interacts with NMYC and CMYC (two members of the oncogene Myc family), and other transcription factors containing a Zip, HLH, or HLH-Zip motif. The NMI protein also interacts with all STATs except STAT2 and augments STAT-mediated transcription in response to cytokines IL2 and IFN-gamma. The NMI mRNA has low expression levels in all human fetal and adult tissues tested except brain and has high expression in cancer cell line-myeloid leukemias. [provided by RefSeq
Sequence: MEADKDDTQQILKEHSPDEFIKDEQNKGLIDEITKKNIQLKKEIQKLETELQEATKEFQIKEDIPETKMKFLSVETPENDSQLSNISCSFQVSSKVPYEISpecifications
| N-myc (and STAT) interactor | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC001268 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 9111 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 9B8 | |
| Mouse monoclonal antibody raised against a partial recombinant NMI. | |
| NMI | |
| NMI | |
| NMI (AAH01268, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Signal Transduction | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.