missing translation for 'onlineSavingsMsg'
Learn More
Learn More
nemo-like kinase, Mouse, Clone: 2B11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant NLK.
Brand: Abnova H00051701-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: DEGRLRYHTCMCKCCFSTSTGRVYTSDFEPVTNPKFDDTFEKNLSSVRQVKEIIHQFILEQQKGNRVPLCINPQSAAFKSFISSTVAQPSEMPPSPLVWESpecifications
| nemo-like kinase | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC064663 | |
| NLK | |
| NLK (AAH64663, 416 a.a. ∼ 515 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Kinases | |
| Primary | |
| Human, Mouse | |
| Antibody |
| ELISA, In situ PLA, Western Blot | |
| 2B11 | |
| Mouse monoclonal antibody raised against a partial recombinant NLK. | |
| NLK | |
| DKFZp761G1211/FLJ21033 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 51701 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction