Learn More
anti-NLGN4Y, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00022829-AP61.1Set
Product Code. 16123244
  • £729.00 / 1 set
Includes: Capture antibody: mouse monoclonal anti-NLGN4Y (100μg) , Detection antibody: rabbit MaxPab affinity purified polyclonal anti-NLGN4Y (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
NLGN4Y
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human NLGN4Y.
KIAA0951
Mouse, Rabbit
RUO
22829
Human
Liquid
ELISA
Unconjugated
NLGN4Y
NLGN4Y
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-NLGN4Y (100 μg) 2. Detection antibody: rabbit MaxPab™ affinity purified polyclonal anti-NLGN4Y (50 μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.