Learn More
anti-NLGN4Y, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00022829-PW1.1Set
Product Code. 16093149
  • £729.00 / 1 set
Includes: Antibody pair for IP: mouse monoclonal anti-NLGN4Y (300μg), Antibody pair for WB: rabbit polyclonal anti-NLGN4Y (50μL)
Specifications
NLGN4Y
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
KIAA0951
Mouse, Rabbit
RUO
22829
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
NLGN4Y
NLGN4Y
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-NLGN4Y (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-NLGN4Y (50 ul)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.