Learn More
Nkx6.1 Mouse, Alexa Fluor 647, Clone: R11-560, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 563338
Description
The R11-560 monoclonal antibody specifically binds to human and mouse homeobox protein Nkx6.1. The transcription factor Nkx6.1 plays a role in pancreatic islet beta cell development through the regulation of HNF1a and insulin genes. Nkx6.1 is considered to be a highly specific transcription factor marking pancreatic beta islet cells, where it is expressed in both developing and mature cells. In addition, Nkx6.1 functions as a transcriptional repressor induced by the Sonic Hedgehog (Shh) signaling pathway in fate determination of several neuronal cell types. During antibody development, the purified R11-560 monoclonal antibody was found to detect Nkx6.1 by western blot and indirect immunofluorescent staining followed by flow cytometric analysis or imaging analysis.
Specifications
| Nkx6.1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| Human Nkx6.1 Recombinant Protein | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| R11-560 | |
| Nkx6.1, NKX6-1, NKX6A, Nkx6-1, Nkx6.1, Nkx6a | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human, Mouse | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.