Learn More
NKX2-5, Mouse, Clone: S1, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant NKX2-5.
Brand: Abnova H00001482-M02.100ug
Description
Homeobox-containing genes play critical roles in regulating tissue-specific gene expression essential for tissue differentiation, as well as determining the temporal and spatial patterns of development (Shiojima et al., 1995 [PubMed 7665173]). It has been demonstrated that a Drosophila homeobox-containing gene called 'tinman' is expressed in the developing dorsal vessel and in the equivalent of the vertebrate heart. Mutations in tinman result in loss of heart formation in the embryo, suggesting that tinman is essential for Drosophila heart formation. Furthermore, abundant expression of Csx, the presumptive mouse homolog of tinman, is observed only in the heart from the time of cardiac differentiation. CSX, the human homolog of murine Csx, has a homeodomain sequence identical to that of Csx and is expressed only in the heart, again suggesting that CSX plays an important role in human heart formation.[supplied by OMIM
Sequence: MFPSPALTPTPFSVKDILNLEQQQRSLAAAGELSARLEATLAPSSCMLAAFKPEAYAGPEAAAPGLPELRAELGRAPSPAKCASAFPAAPAFYPRAYSDPDPAKDPRAEKKELCALQKAVELEKTEADNAERPRARRRRKPRVLFSQAQVYELERRFKQQRYLSAPERDQLASVLKLTSTQVKIWFQNRRYKCKRQRQDQTLELVGLPPPPPPPARRIAVPVLVRDGKPCLGDSAPYAPAYGVGLNPYGYNAYPAYPGYGGAACSPGYSCTAAYPAGPSPAQPATAAANNNFVNFGVGDLNAVQSPGIPQSNSGVSTLHGIRAWSpecifications
| NKX2-5 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC025711 | |
| NKX2-5 | |
| NKX2-5 (AAH25711, 1 a.a. ∼ 324 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| S1 | |
| Mouse monoclonal antibody raised against a full length recombinant NKX2-5. | |
| NKX2-5 | |
| CHNG5/CSX/CSX1/NKX2.5/NKX2E/NKX4-1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1482 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.