missing translation for 'onlineSavingsMsg'
Learn More

anti-NKp46/NCR1 FITC, Clone: 29A1.4, Invitrogen™

NKp46/NCR1 Monoclonal antibody A14752 from Invitrogen specifically detects NKp46/NCR1 in Mouse samples. It is validated for Flow Cytometry.

Brand:  Invitrogen A14752

Product Code. 13355594

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

NKp46/NCR1 Monoclonal antibody A14752 from Invitrogen specifically detects NKp46/NCR1 in Mouse samples. It is validated for Flow Cytometry.

Cytotoxicity-activating receptor that may contribute to the increased efficiency of activated natural killer (NK) cells to mediate tumor cell lysis.
Host Species: Rat
Cross Reactivity: Mouse
Isotype: IgG2a
Clone: 29A1.4
Immunogen: NKP46-IgG1 Fc fusion protein.
Formulation: 50μg Purified Antibody (0.5mg/ml) in proprietary buffer, with 0.09% sodium azide
Synonyms: CD335, LY94, NK-p46, NKP46, NK cell-activating receptor, lymphocyte antigen 94 homolog activating NK-receptor, NK-p46, natural killer cell p46-related protein
Uniprot Accession: Q8C567
Entrez GeneID: 17086
Gene Symbol: Ncr1
This product detects NKp46/NCR1 in Mouse samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is NKP46-IgG1 Fc fusion protein.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NKp46/NCR1
Monoclonal
FITC
Q8C567
Ncr1
NKP46-IgG1 Fc fusion protein.
Primary
Mouse
Purified
Flow Cytometry
29A1.4
50μg Purified Antibody (0.5mg/ml) in proprietary buffer, with 0.09% sodium azide
CD335, LY94, NK-p46, NKP46, NK cell-activating receptor, lymphocyte antigen 94 homolog activating NK-receptor, NK-p46, natural killer cell p46-related protein
Rat
50 μg
17086
Store at 4° C
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.