Learn More
NKp46 Mouse anti-Human, Unlabeled, Clone: 9E2/NKP46, BD
Mouse Monoclonal Antibody
£113.93 - £371.93
Specifications
| Antigen | NKp46 |
|---|---|
| Clone | 9E2/NKP46 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15809108
|
BD Biosciences
557911 |
0.1 mg |
£347.00 £113.93 / Each Save £233.07 67% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15809078
|
BD Biosciences
557847 |
0.5 mg |
£387.00 £371.93 / Each Save £15.07 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NKp46, a major cytoxicity receptor of NK cells, is a type I membrane glycoprotein (46kDaa) is expressed on resting and activated NK cells. Its extracellular region contains two C2-type, Ig-like domains. The transmembrane domain contains a positively charged amino acid (Arg) which could be involved in stabilizing the association with CD3ζ. Its intracellular region does not contain immunoreceptor tyrosine-based activating motifs (ITAM), but it is linked to intracytoplasmic transduction machinery by its association with CD3ζ and FcεRIγ adaptor proteins. NKp46 along with NKp30 and NKp44 are referred to as natural cytoxicity receptors (NCR) and play a very important role in killing virus-infected, tumor cells and MHC-class I-unprotected cells. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: 9E2
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
| NKp46 | |
| Flow Cytometry | |
| Unconjugated | |
| RUO | |
| Human | |
| IgG1 κ | |
| Affinity Purified |
| 9E2/NKP46 | |
| Monoclonal | |
| Mouse | |
| Immunology | |
| Human NKp46 Recombinant Protein | |
| Primary | |
| Store undiluted at 4°C |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.