missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHE3 Mouse anti-Rabbit, Rat, Clone: 19F5, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA146355
This item is not returnable.
View return policy
Description
This antibody has 83% sequence homology with mouse. Does not react with human samples. Suggested positive control: rat kidney membrane.
NHE3 (also known as SLC9A3 - Sodium/hydrogen exchanger 3) is a member of the sodium/hydrogen exchanger family that plays a crucial role in acid-base and volume homeostasis by mediating the majority of sodium and bicarbonate reabsorption in the proximal tubule of the kidney. Mutations in the gene can lead to Diarrhea 8.
Specifications
| NHE3 | |
| Monoclonal | |
| 1 mg/mL | |
| Slc9a3 | |
| Sodium/hydrogen exchanger 3 antibody, Na (+)/H(+) exchanger 3 antibody, NHE-3 antibody, Solute carrier family 9 member 3 antibody, SLC9A3 antibody | |
| Mouse | |
| Protein G | |
| RUO | |
| 100009430, 24784 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| 19F5 | |
| Unconjugated | |
| P26432, P26433 | |
| SLC9A3 | |
| Fusion protein representing the C-terminal 131 amino acids of rabbit NHE3. | |
| 100 μL | |
| Primary | |
| Rabbit, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction