missing translation for 'onlineSavingsMsg'
Learn More

NHE-1 (Na+/H+ Exchangers) Mouse, Unlabeled, Clone: 54, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 611775

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15806029

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

The extrusion of H+ in exchange for extracellular Na+ is important for many cellular processes, such as pH homeostasis, volume regulation, and transepithelial ion and water transport. Na+/H+ Exchangers (NHE) are integral membrane proteins that mediate electroneutral exchange of one Na+ ion for one H+ ion. Six NHE forms, NHE-1 thru -6, have been identified. NHE-1 and NHE-6 are widely expressed, while the other NHE forms have restricted expression. The common structure of all NHE forms includes 10-12 N-terminal membrane (M) spanning regions, a conserved M6 and M7 region that may participate in ion transport, and a large C-terminal cytoplasmic region that may be involved in the regulation of ion exchange activity. NHE-1, for example, contains 12 M regions plus domains for volume sensitivity, calmodulin-binding, CHP-binding, and PKC phosphorylation in the cytoplasmic region. Regulation of NHE-1 ion exchange activity may occur through phosphoinositide binding, as well as PKC- and PKA-dependent signaling pathways. Mutation of NHE-1 in mice causes neuronal death in the cerebellum and brainstem, leading to ataxia and seizures. Thus, NHE-1 is a ubiquitous NHE that is essential for normal brain function.

Although this antibody was developed against the NHE-1 antigen, investigators should note that crossreactivity to other NHE isoforms or variants may be possible.

Although this antibody was developed against the NHE-1 antigen, investigators should note that crossreactivity to other NHE isoforms or variants may be possible.

Specifications

Specifications

NHE-1 (Na+/H+ Exchangers)
Monoclonal
250μg/mL
Na+/H+ Exchangers
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
54
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Rat NHE-1 aa. 682-801
150 μg
Cell Biology
Canine, Human, Mouse, Rat
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.