Learn More
nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, beta, Mouse, Clone: 1B5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant NFKBIB.
Brand: Abnova H00004793-M01.100ug
Description
NFKB1 (MIM 164011) or NFKB2 (MIM 164012) is bound to REL (MIM 164910), RELA (MIM 164014), or RELB (MIM 604758) to form the NFKB complex. The NFKB complex is inhibited by I-kappa-B proteins (NFKBIA, MIM 164008, or NFKBIB), which inactivate NF-kappa-B by trapping it in the cytoplasm. Phosphorylation of serine residues on the I-kappa-B proteins by kinases (IKBKA, MIM 600664 or IKBKB, MIM 603258) marks them for destruction via the ubiquitination pathway, thereby allowing activation of the NF-kappa-B complex. Activated NFKB complex translocates into the nucleus and binds DNA at kappa-B-binding motifs such as 5-prime GGGRNNYYCC 3-prime or 5-prime HGGARNYYCC 3-prime (where H is A, C, or T; R is an A or G purine; and Y is a C or T pyrimidine).[supplied by OMIM
Sequence: EDGDTALHLAVIHQHEPFLDFLLGFSAGTEYMDLQNDLGQTALHLAAILGETSTVEKLYAAGAGLCVAERRGHTALHLACRVGAHACARASpecifications
| nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, beta | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC015528 | |
| NFKBIB | |
| NFKBIB (AAH15528, 56 a.a. ∼ 145 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (PFA fixed) | |
| 1B5 | |
| Mouse monoclonal antibody raised against a partial recombinant NFKBIB. | |
| NFKBIB | |
| IKBB/TRIP9 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 4793 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.