Learn More
neurogenic differentiation 1, Mouse, Clone: 3D11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant NEUROD1.
Brand: Abnova H00004760-M02.100ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the NeuroD family of basic helix-loop-helix (bHLH) transcription factors. The protein forms heterodimers with other bHLH proteins and activates transcription of genes that contain a specific DNA sequence known as the E-box. It regulates expression of the insulin gene, and mutations in this gene result in type II diabetes mellitus. [provided by RefSeq
Sequence: QDMPPHLPTASASFPVHPYSYQSPGLPSPPYGTMDSSHVFHVKPPPHAYSAALEPFFESPLTDCTSPSFDGPLSPPLSINGNFSFKHEPSAEFEKNYAFTSpecifications
neurogenic differentiation 1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC009046 | |
NEUROD1 | |
NEUROD1 (AAH09046, 201 a.a. ∼ 300 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Neurobiology | |
Primary | |
Human | |
Antibody |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
3D11 | |
Mouse monoclonal antibody raised against a partial recombinant NEUROD1. | |
NEUROD1 | |
BETA2/BHF-1/NEUROD/bHLHa3 | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
4760 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |