Learn More
NeuroD1, clone: 6E9G12, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517137
Description
This gene encodes a member of the NeuroD family of basic helix-loop-helix (bHLH) transcription factors. The protein forms heterodimers with other bHLH proteins and activates transcription of genes that contain a specific DNA sequence known as the E-box. It regulates expression of the insulin gene, and mutations in this gene result in type II diabetes mellitus.
MA5-17137 targets NeuroD in pep-ELISA and WB applications and shows reactivity with Human and Mouse samples.The MA5-17137 immunogen is purified recombinant fragment of human Neurod (amino acids: 26-91) expressed in E. Coli.MA5-17137 detects NeuroD which has a predicted molecular weight of approximately 40kDa.
Specifications
| NeuroD1 | |
| Monoclonal | |
| 1mg/mL | |
| PBS with 0.5% proprietary stabilizer and 0.05% sodium azide | |
| Q13562, Q60867 | |
| Mouse | |
| Purified | |
| RUO | |
| 18012, 4760 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 6E9G12 | |
| Unconjugated | |
| NEUROD1 | |
| NEUROD1 | |
| Purified recombinant fragment of human Neurod (amino acids: 26-91) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.