missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFV2, Mouse, Clone: 1A10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant NDUFV2.
Brand: Abnova H00004729-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: EAIQKKLGIKVGETTPDKLFTLIEVECLGACVNAPMVQINDNYYEDLTAKDIEEIIDELKAGKIPKPGPRSGRFSCEPAGGLTSLTEPPKGPGFGVQAGLSpecifications
| NDUFV2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_021074 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4729 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| ELISA, Western Blot | |
| 1A10 | |
| Mouse monoclonal antibody raised against a partial recombinant NDUFV2. | |
| NDUFV2 | |
| NDUFV2 | |
| NDUFV2 (NP_066552, 150 a.a. ∼ 249 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction