missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFB8 Monoclonal Antibody (20E9DH10C12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ 459210
This item is not returnable.
View return policy
Description
Positive control: Isolated mitochondria from Human heart, bovine heart, rat heart and mouse heart. Skeletal muscle tissue.
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. [UniProt].
Specifications
| NDUFB8 | |
| Monoclonal | |
| 1 mg/mL | |
| HEPES buffered saline with 0.02% sodium azide | |
| O95169, Q02372, Q9D6J5 | |
| NDUFB8 | |
| Bovine Complex I. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Frozen), Western Blot | |
| 20E9DH10C12 | |
| Unconjugated | |
| NDUFB8 | |
| 19 kDa; 2900010I05Rik; AI987932; ASHI; CI-ASHI; complex I ASHI subunit; complex I-ASHI; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) 1 beta subcomplex 8; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 8, 19kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 8, mitochondrial; NADH dehydrogenase 1 beta subcomplex 8; NADH:ubiquinone oxidoreductase ASHI subunit; NADH:ubiquinone oxidoreductase subunit B8; NADH-ubiquinone oxidoreductase ASHI subunit; NDUFB8 | |
| Mouse | |
| Purified | |
| RUO | |
| 282517, 293991, 4714, 67264 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction