Learn More
NDUFA1, Mouse, Clone: 2A4, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant NDUFA1.
Brand: Abnova H00004694-M10.100ug
Description
The human NDUFA1 gene codes for an essential component of complex I of the respiratory chain, which transfers electrons from NADH to ubiquinone. It has been noted that the N-terminal hydrophobic domain has the potential to be folded into an alpha-helix spanning the inner mitochondrial membrane with a C-terminal hydrophilic domain interacting with globular subunits of complex I. The highly conserved two-domain structure suggests that this feature is critical for the protein function and might act as an anchor for the NADH:ubiquinone oxidoreductase complex at the inner mitochondrial membrane. However, the NDUFA1 peptide is one of about 31 components of the hydrophobic protein (HP) fraction of complex I which is involved in proton translocation. Thus the NDUFA1 peptide may also participate in that function. [provided by RefSeq]
Sequence: MWFEILPGLSVMGVCLLIPGLATAYIHRFTNGGKEKRVAHFGYHWSLMERDRRISGVDRYYVSKGLENIDSpecifications
| NDUFA1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC000266 | |
| NDUFA1 | |
| NDUFA1 (AAH00266, 1 a.a. ∼ 70 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA | |
| 2A4 | |
| Mouse monoclonal antibody raised against a full-length recombinant NDUFA1. | |
| NDUFA1 | |
| CI-MWFE/MWFE/ZNF183 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4694 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.