Learn More
Invitrogen™ NCOA5 Monoclonal Antibody (1E9)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522408
Description
Peptide Sequence: MAGMPGTAET FETPETCGTT DMRDSRDPMY RREGSYDRYL RMDDYCRRKD DSYFDRYRDS FDGRGPPGPE SQSRAKERLK REERRREELY RQYFEEIQRR FDAERPVDCS VIVVNKQTKD YAESVGRKVR DLGMVVDLIF LNTEVSLSQA LEDVSRGGSP FAIVITQQHQ IHRSCTVNIM FGTPQEHRNM PQADAMVLVA RNYERYKNEC REKEREEIAR QAAKMADEAI LQERERGGPE EGVRGGHPPA IQSLINLLAD NRYLTAEETD KIINYLRERK ERLMRSSTDS LPGELRGRAE ARFPANHSGR PRVPR.
This gene encodes a coregulator for the alpha and beta estrogen receptors and the orphan nuclear receptor NR1D2. The protein localizes to the nucleus, and is thought to have both coactivator and corepressor functions. Its interaction with nuclear receptors is independent of the AF2 domain on the receptors, which is known to regulate interaction with other coreceptors. Several alternatively spliced transcript variants have been found for this gene.
Specifications
| NCOA5 | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9HCD5 | |
| NCOA5 | |
| NCOA5 (AAH56872, 1 a.a. ~ 315 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA | |
| 1E9 | |
| Unconjugated | |
| NCOA5 | |
| bA465L10.6; CIA; coactivator independent of AF-2; KIAA1637; NCOA5; NCoA-5; Nuclear receptor coactivator 5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 57727 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.