missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NCAM-1/CD56 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
£188.00 - £427.00
Specifications
| Antigen | NCAM-1/CD56 |
|---|---|
| Clone | 301040 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18731679
![]() |
R&D Systems
FAB2408P-025 |
25 Tests |
£188.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18729716
![]() |
R&D Systems
FAB2408P-100 |
100 Tests |
£427.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NCAM-1/CD56 Monoclonal specifically detects NCAM-1/CD56 in Human samples. It is validated for Flow Cytometry.Specifications
| NCAM-1/CD56 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 4684 | |
| Mouse myeloma cell line NS0-derived recombinant human NCAM-1/CD56 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| 301040 | |
| Flow Cytometry | |
| R-PE | |
| Mouse | |
| Human | |
| CD56, CD56/ NCAM-1, CD56 antigen, MSK39, NCAM1, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM | |
| NCAM1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human NCAM-1/CD56 in ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ALCAM, rhBCAM, rhEPCAM, rhMCAM, rhNCAM-1-L1, recombinant mouse (rm) MAdCAM-1, rmNCAM-1-L1, or rmOCAM is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title