Learn More
NBS1 Mouse, Unlabeled, Clone: 34, BD
Mouse Monoclonal Antibody
£343.55
Specifications
| Antigen | NBS1 |
|---|---|
| Clone | 34 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15836049
|
BD Biosciences
611871 |
150 μg | |||||||
Description
Nijmegen Breakage Syndrome (NBS) is characterized by extreme radiation sensitivity and chromosomal instability. The NBS1 gene product, p95/nibrin/NBS1, forms a complex with Rad50 and Mre11. Cells deficient in this complex have problems with DNA double-strand break repair, cell cycle checkpoint control, and telomere length maintenance. NBS1 contains a forkhead-associated domain (FHA) and a breast cancer carboxy-terminal domain (BRCT) in the N-terminal region. Both of these domains have been found in DNA-damage responsive cell cycle checkpoint proteins. The complex containing NBS1, Rad50, and Mre11 possesses manganese-dependent single stranded DNA endonuclease and 3′ to 5′ exonuclease activities. In addition, NBS1 is required for DNA-damage dependent phosphorylation of Mre11. This phosphorylation may be required for proper nuclear localization of the NBS1-Rad50-Mre11 complex to sites of DNA double-strand breaks. NBS1 interacts directly with telomere repeat binding factor, TRF1, via its C-terminal region, and both NBS1 and Mre11 co-localize with TRF1 at promyelocytic leukemia nuclear bodies. Thus, the NBS1-Rad50-Mre11 complex may be important for both DNA damage repair, and telomere length maintenance.
Host Species: Mouse
Clone: 34
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human NBS1 aa. 620-732
Immunofluorescence, Western Blotting
Specifications
| NBS1 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified | |
| Nijmegen Breakage Syndrome-1 |
| 34 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Chicken, Human, Mouse, Rat | |
| Human NBS1 aa. 620-732 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.