missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asparagine synthetase domain containing 1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human NBLA00058 protein.
Brand: Abnova H00054529-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MCGICCSVNFSAEHFSQDLKEDLLYNLKQRGPNSSKQLLKSDVNYQCLFSAHVLHLRGVLTTQPVEDERGNVFLWNGEIFSGIKVEAEENDTQILFNYLSSCKNESEILSLFSEVQGPWSFIYYQASSHYLWFGRDFFGRRSLLWHFSNLGKSFCLSSVGTQTSGLANQWQEVPASGLFRIDLKSTVISRCIILQLYPWKYISRENIIEENVNSLSQISADLPAFVSVVANEAKLYLEKPVVPLNMMLPQAALETHCSNISNVPPTREILQVFLTDVHMKEVIQQFIDVLSVAVKKRVLCLPRDENLTANEVLKTCDRKANVAILFSGGIDSMVIATLADRHIPLDEPIDLLNVAFIAEEKTMPTTFNREGNKQKNKCEIPSEEFSKDVAAAAADSPNKHVSVPDRITGRAGLKELQAVSPSRIWNFVEINVSMEELQKLRRTRICHLIRPLDTVLDDSIGCAVWFASRGIGWLVAQEGVKSYQSNAKVVLTGIGADEQLAGYSRHRVRFQSHGLEGLNKEIMMELGRISSRNLGRDDRVIGDHGKEARFPFLDENVVSFLNSLPIWEKANLTLPRGIGEKLLLRLAAVELGLTASALLPKRAMQFGSRIAKMEKINEKASDKCGRLQIMSLENLSIEKETKLSpecifications
| asparagine synthetase domain containing 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human NBLA00058 protein. | |
| ASNSD1 | |
| FLJ20752/NBLA00058/NS3TP1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 54529 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_019048.1 | |
| ASNSD1 | |
| NBLA00058 (NP_061921.1, 1 a.a. ∼ 643 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction