Learn More
nuclear prelamin A recognition factor, Mouse, Clone: 7D9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant NARF.
Brand: Abnova H00026502-M03A.200uL
Description
Several proteins have been found to be prenylated and methylated at their carboxyl-terminal ends. Prenylation was initially believed to be important only for membrane attachment. However, another role for prenylation appears to be its importance in protein-protein interactions. The only nuclear proteins known to be prenylated in mammalian cells are prelamin A- and B-type lamins. Prelamin A is farnesylated and carboxymethylated on the cysteine residue of a carboxyl-terminal CaaX motif. This post-translationally modified cysteine residue is removed from prelamin A when it is endoproteolytically processed into mature lamin A. The protein encoded by this gene binds to the prenylated prelamin A carboxyl-terminal tail domain. It may be a component of a prelamin A endoprotease complex. The encoded protein is located in the nucleus, where it partially colocalizes with the nuclear lamina. It shares limited sequence similarity with iron-only bacterial hydrogenases. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene, including one with a novel exon that is generated by RNA editing. [provided by RefSeq
Sequence: MKCEHCTRKECSKKTKTDDQENVSADAPSPAQENGEKGEFHKLADAKIFLSDCLACDSCMTAEEGVQLSQQNAKDFFRVLNLNKKCDTSKHKVLVVSVCPSpecifications
| nuclear prelamin A recognition factor | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_031968 | |
| NARF | |
| NARF (NP_114174, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
| ELISA, Western Blot | |
| 7D9 | |
| Mouse monoclonal antibody raised against a partial recombinant NARF. | |
| NARF | |
| DKFZp434G0420/FLJ10067/IOP2 | |
| Mouse | |
| 200 μL | |
| Yes | |
| 26502 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.