Learn More
Nanog Mouse anti-Mouse, Alexa Fluor 488, Clone: M55-312, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560278
Description
The M55-312 monoclonal antibody reacts with mouse Nanog (named for Tir Na Nog, the land of the ever-young of Celtic mythology), which is a homeobox transcription factor required for the maintenance of the undifferentiated state of pluripotent stem cells. Nanog expression counteracts the differentiation-promoting signals induced by the extrinsic factors LIF (Leukemia Inhibitory Factor) and BMP (Bone Morphogenic Protein). When Nanog expression is down-regulated, cell differentiation can proceed. Proteins that regulate Nanog expression include transcription factors Oct4, SOX2, FoxD3, and Tcf3 and tumor suppressor p53.
Specifications
| Nanog | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| M55-312 | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| Mouse Nanog Recombinant Protein | |
| 50 Tests | |
| Cell Biology | |
| Mouse | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.