Learn More
anti-N Cadherin, Polyclonal, Abcam™
This product is for research use only and not for diagnostic or therapeutic procedures. Customer is granted a limited, non-exclusive license for internal research and shall not attempt to reverse engineer.
Brand: Abcam AB18203-100UG
Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Mouse, Rat, Human, Xenopus laevis. Predicted to work with Chicken, Cow
Immunogen: Synthetic peptide conjugated to KLH derived from within residues 800 - 900 of Human N Cadherin
Applications:
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blotting
Specifications
| N-Cadherin | |
| 0.6-1mg/mL | |
| Rabbit | |
| Affinity Purified | |
| Cancer and Tumor Biology, Signal Transduction, Stem Cell Biology | |
| Bovine, Chicken, Human, Mouse, Rat, Xenopus species |
| Polyclonal | |
| Unconjugated | |
| Synthetic peptide conjugated to KLH derived from within residues 800 - 900 of Human N Cadherin | |
| 100 μg | |
| Primary | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.