missing translation for 'onlineSavingsMsg'
Learn More

N-Cadherin Mouse, Unlabeled, Clone: 32, BD

Mouse Monoclonal Antibody

£188.32 - £284.36

Specifications

Antigen N-Cadherin
Clone 32
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15815709
View Documents
BD Biosciences
610920
50 μg
ONLINE EXCLUSIVE PRICE
£196.00
£188.32 / 1 set
Save £7.68
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15825709
View Documents
BD Biosciences
610921
150 μg
ONLINE EXCLUSIVE PRICE
£297.00
£284.36 / 1 set
Save £12.64
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cadherins are a family of Ca2+;-dependent intercellular adhesion molecules that play a central role in controlling morphogenetic movements during development. Their function is regulated by association with the actin cytoskeleton by a complex of cytoplasmic proteins called the catenins (α, β, γ). Members of the cadherin family include P-cadherin, E-cadherin (uvomorulin), N-cadherin (neural cadherin), R-cadherin, cadherin 5, L-CAM, and EP-cadherin. N-cadherin mRNA is found at elevated levels in brain and heart and at a much lower level in liver. Mechanisms such as mRNA expression, cytokine modulation, and protease-mediated turnover modulate N-cadherin protein levels during development. In addition, N-cadherin function is indirectly regulated by endogenous kinases and phosphatases. Tyrosine phosphorylation of β-catenin complexed with N-cadherin results in dissociation of N-cadherin from actin. However, N-cadherin also interacts with a PTP1B-like phosphatase that dephosphorylates β-catenin and promotes N-cadherin/actin association. Thus, N-cadherin is an integral adhesion molecule whose function is regulated by protein-protein interactions and phosphorylation/dephosphorylation events.

Immunofluorescence, Western Blotting

Specifications

Specifications

N-Cadherin
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
32
Western Blot
Unconjugated
RUO
Chicken, Human, Mouse, Rat
Mouse N-Cadherin aa. 802-819
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.