Learn More
myelin transcription factor 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MYT1.
Brand: Abnova H00004661-A01.50uL
Description
The protein encoded by this gene is a member of a family of neural specific, zinc finger-containing DNA-binding proteins. The protein binds to the promoter regions of proteolipid proteins of the central nervous system and plays a role in the developing nervous system. [provided by RefSeq
Sequence: DYASFDAQVFGKRMLAPKIQTSETSPKAFQCFDYSQDAEAAHMAATAILNLSTRCWEMPENLSTKPQDLPSKSVDIEVDENGTLDLSMHKHRKRENAFPSSpecifications
| myelin transcription factor 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant MYT1. | |
| MYT1 | |
| C20orf36/MTF1/MYTI/PLPB1 | |
| Mouse | |
| 50 μL | |
| Kinases | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_004535 | |
| MYT1 | |
| MYT1 (NP_004526, 586 a.a. ∼ 685 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 4661 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.