missing translation for 'onlineSavingsMsg'
Learn More

MYPT1 Mouse, Unlabeled, Clone: 20, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 612164

 View more versions of this product

Product Code. 15866119

  • ONLINE EXCLUSIVE PRICE
    £198.00
    £190.20 / 1 set
    Save £7.80
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Protein phosphatase-1 (PP1) is a major serine/threonine phosphatase that is involved in various cellular processes in eukaryotic cells. The PP1 catalytic subunit may be directed to particular subcellular locations by targeting subunits, which can modify the substrate specificity of the enzyme. The PP1 enzyme associated with the myofibrils of striated and smooth muscle forms a complex with Myosin Phosphatase Targeting subunit 1 (MYPT1). MYPT1 contains seven ankyrin repeats in the N-terminal region, and a leucine zipper (LZ) motif in the C-terminus. Alternative splicing of the MYPT1 mRNA leads to expression of a truncated MYPT1 that lacks the leucine zipper region. The expression of MYPT1 splice variants correlate with a smooth muscle phenotype. In the tonic contracting chicken aorta, only the full length MYPT1 is expressed, while in the phasic contracting chicken gizzard, only the truncated MYPT1 is expressed. The leucine zipper motif may be required for association with cGMP-dependent protein kinase I. Thus, MYPT1 may direct not only PP1 enzymatic activity toward myosin, but also may regulate the cGMP responsiveness of PP1 through expression of alternative splice variants.

Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

MYPT1
Monoclonal
250μg/mL
Myosin Phosphatase Targeting subunit 1
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
20
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Rat MYPT1 aa. 723-840
50 μg
Cell Biology
Human, Mouse, Rat
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.