Learn More
myogenic differentiation 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MYOD1.
Brand: Abnova H00004654-A01.50uL
Description
This gene encodes a nuclear protein that belongs to the basic helix-loop-helix family of transcription factors and the myogenic factors subfamily. It regulates muscle cell differentiation by inducing cell cycle arrest, a prerequisite for myogenic initiation. The protein is also involved in muscle regeneration. It activates its own transcription which may stabilize commitment to myogenesis. [provided by RefSeq
Sequence: MDYSGPPSGARRRNCYEGAYYNEAPSEPRPGKSAAVSSLDCLSSIVERISTESPAAPALLLADVPSESPPRRQEAAAPSEGESSGDPTQSPDAAPQCPAGANPNPIYQVLSpecifications
| myogenic differentiation 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant MYOD1. | |
| MYOD1 | |
| MYF3/MYOD/PUM/bHLHc1 | |
| Mouse | |
| 50 μL | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_002478 | |
| MYOD1 | |
| MYOD1 (NP_002469, 211 a.a. ∼ 320 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 4654 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.