Learn More
anti-MYOC Matched Antibody Pair, Abnova
Brand: Abnova H00004653-AP11.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-MYOC (100μg); Mouse monoclonal anti-MYOC, IgG1κ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
MYOC encodes the protein myocilin, which is believed to have a role in cytoskeletal function. MYOC is expressed in many occular tissues, including the trabecular meshwork, and was revealed to be the trabecular meshwork glucocorticoid-inducible response protein (TIGR). The trabecular meshwork is a specialized eye tissue essential in regulating intraocular pressure, and mutations in MYOC have been identified as the cause of hereditary juvenile-onset open-angle glaucoma.
ELISA
Specifications
| MYOC | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human MYOC. | |
| GLC1A/GPOA/JOAG/JOAG1/TIGR/myocilin | |
| Mouse, Rabbit | |
| 1 Set | |
| Membrane Receptors | |
| 4653 | |
| Human | |
| IgG1 |
| ELISA | |
| Unconjugated | |
| MYOC | |
| MYOC | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-MYOC (100 μg) 2. Detection antibody: mouse monoclonal anti-MYOC, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.