Learn More
MYO1B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522347
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T. Predicted reactivity: Mouse (100%), Rat (100%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MYO1B Polyclonal antibody specifically detects MYO1B in Human samples. It is validated for Western Blot.Specifications
MYO1B | |
Polyclonal | |
Unconjugated | |
MYO1B | |
AA406997; MIH-L; MMIa; MMI-alpha; molecular motor; MYH-1c; Myo1a; MYO1B; MYO1B variant protein; myosin heavy chain myr 1; Myosin I alpha; Myosin IA (Myosin I alpha) (MMI-alpha) (MMIa) (MIH-L); myosin Ib; myosin-I alpha; myosin-Ib; myr1; Unconventional myosin-Ib | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4430 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.91 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O43795 | |
MYO1B | |
Recombinant fragment corresponding to a region within amino acids 916 and 1136 of MYO1B (Uniprot ID#O43795) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |