missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Myeloperoxidase Monoclonal Antibody (2C7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA180878
This item is not returnable.
View return policy
Description
This antibody does not react with rat. Non-reducing conditions are recommended for Western blot applications. A suggested positive control for immunohistochemical applications is bone marrow. Mouse anti Human myeloperoxidase antibody, clone 2C7 recognizes human myeloperoxidase (MPO).
Myeloperoxidase (MPO) is a heme protein that is synthesized during myeloid differentiation and is a major component of neutrophil azurophilic granules. It is produced as a single chain precursor and subsequently cleaved into a light and heavy chain. The mature MPO is a tetramer composed of 2 light chains and 2 heavy chains. This enzyme is responsible for producing hypohalous acids that are central to the microbicidal activity of neutrophils. Diseases that are associated with MPO include Myeloperoxidase Deficiency and Alzheimer's Disease, Familial, 1.
Specifications
| Myeloperoxidase | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P05164 | |
| MPO | |
| Human Myeloperoxidase. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2C7 | |
| Unconjugated | |
| MPO | |
| 84 kDa myeloperoxidase; 89 kDa myeloperoxidase; mKIAA4033; MPO; Myeloperoxidase; Myeloperoxidase heavy chain; Myeloperoxidase light chain | |
| Mouse | |
| Protein A | |
| RUO | |
| 4353 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu