Learn More
v-myb myeloblastosis viral oncogene homolog (avian)-like 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human MYBL2 protein.
Brand: Abnova H00004605-D01P.100ug
Description
The protein encoded by this gene, a member of the MYB family of transcription factor genes, is a nuclear protein involved in cell cycle progression. The encoded protein is phosphorylated by cyclin A/cyclin-dependent kinase 2 during the S-phase of the cell cycle and possesses both activator and repressor activities. It has been shown to activate the cell division cycle 2, cyclin D1, and insulin-like growth factor-binding protein 5 genes. Transcript variants may exist for this gene, but their full-length natures have not been determined. [provided by RefSeq
Sequence: MSRRTRCEDLDELHYQDTDSDVPEQRDSKCKVKWTHEEDEQLRALVRQFGQQDWKFLASHFPNRTDQQCQYRWLRVLNPDLVKGPWTKEEDQKVIELVKKYGTKQWTLIAKHLKGRLGKQCRERWHNHLNPEVKKSCWTEEEDRIICEAHKVLGNRWAEIAKMLPGRTDNAVKNHWNSTIKRKVDTGGFLSESKDCKPPVYLLLELEDKDGLQSAQPTEGQGSLLTNWPSVPPTIKEEENSEEELAAATTSKEQEPIGTDLDAVRTPEPLEEFPKREDQEGSPPETSLPYKWVVEAANLLIPAVGSSLSEALDLIESDPDAWCDLSKFDLPEEPSAEDSINNSLVQLQASHQQQVLPPRQPSALVPSVTEYRLDGHTISDLSRSSRGELIPISPSTEVGGSGIGTPPSVLKRQRKRRVALSPVTENSTSLSFLDSCNSLTPKSTPVKTLPFSPSQFLNFWNKQDTLELESPSLTSTPVCSQKVVVTTPLHRDKTPLHQKHAAFVTPDQKYSMDNTPHTPTPFKNALEKYGPLKPLPQTPHLEEDLKEVLRSEAGIELIIEDDIRPEKQKRKPGLRRSPIKKVRKSLALDIVDEDVKLMMSTLPKSLSLPTTAPSNSSSLTLSGIKEDNSLLNQGFLQAKPEKAAVAQKPRSHFTTPAPMSSAWKTVACGGTRDQLFMQEKARQLLGRLKPSHTSRTLILSSpecifications
| v-myb myeloblastosis viral oncogene homolog (avian)-like 2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human MYBL2 protein. | |
| MYBL2 | |
| B-MYB/BMYB/MGC15600 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| In situ PLA, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_002466.2 | |
| MYBL2 | |
| MYBL2 (NP_002457.1, 1 a.a. ∼ 700 a.a) full-length human protein. | |
| 100 μg | |
| Apoptosis | |
| 4605 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.