missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MUC5AC Monoclonal Antibody (45M1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA121907
This item is not returnable.
View return policy
Description
Recommended positive controls: Suitable tissue control sections for Immunohistochemistry:Stomach. Amount of BSA in this product is equal to or less than 0.01%.
The antibody 2-25LE reacts with Lewisb blood group antigen, a carbohydrate determinant carried on both glycolipids and glycoproteins, detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals.
Specifications
| MUC5AC | |
| Monoclonal | |
| Unconjugated | |
| MUC5AC | |
| 2210005L13Rik; gastric mucin; LeB; lewis B blood group antigen; LOW QUALITY PROTEIN: mucin-5AC; major airway glycoprotein; MGM; MUC5; MUC5AC; MUC-5AC; mucin; mucin 5, subtypes A and C; mucin 5, subtypes A and C, tracheobronchial/gastric; mucin 5ac; mucin 5AC, oligomeric mucus/gel-forming; mucin 5AC, oligomeric mucus/gel-forming pseudogene; mucin-5 subtype AC, tracheobronchial; Mucin-5AC; TBM; Tracheobronchial mucin | |
| Mouse | |
| IgG fraction | |
| RUO | |
| 4586 | |
| 4°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 45M1 | |
| PBS with 0.09% sodium azide | |
| P98088 | |
| MUC5AC | |
| Fluid of an ovarian mucinous cyst belonging to an O Le (a-b) patient. | |
| 500 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction