missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-MUC-4, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Specifications
| Host Species | Rabbit |
|---|---|
| Isotype | IgG |
| Immunogen | A genomic peptide made to an internal region of the human MUC4 protein (within residues 1950-2100). [Swiss-Prot Q99102] |
| Antigen | MUC-4 |
| Primary or Secondary | Primary |
Description
MUC4 Polyclonal specifically detects MUC4 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Rabbit | |
| A genomic peptide made to an internal region of the human MUC4 protein (within residues 1950-2100). [Swiss-Prot Q99102] | |
| Primary | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Q99102, Q8JZM8 | |
| PBS, 0.1% BSA, 50% Glycerol with 0.05% Sodium Azide |
| IgG | |
| MUC-4 | |
| RUO | |
| This antibody is specific for the beta chain of MUC4. | |
| 4585 | |
| 1.0 mg/ml |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title