missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTMR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542117
This item is not returnable.
View return policy
Description
Peptide sequence: KEDVYTKTIS LWSYINSQLD EFSNPFFVNY ENHVLYPVAS LSHLELWVNY Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
MTMR1 is a member of the myotubularin related family of proteins. Members of this family contain the consensus sequence for the active site of protein tyrosine phosphatases.This gene encodes a member of the myotubularin related family of proteins. Members of this family contain the consensus sequence for the active site of protein tyrosine phosphatases. Alternatively spliced variants have been described but their biological validity has not been determined.
Specifications
| MTMR1 | |
| Polyclonal | |
| Unconjugated | |
| MTMR1 | |
| AW049210; LOW QUALITY PROTEIN: myotubularin-related protein 1; Mtmr1; myotubularin related protein 1; myotubularin-related protein 1; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase; phosphatidylinositol-3-phosphate phosphatase | |
| Rabbit | |
| Protein A | |
| RUO | |
| 8776 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q13613 | |
| MTMR1 | |
| Synthetic peptide directed towards the C-terminal of human MTMR1 (aa 551-600). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction