Learn More
membrane-spanning 4-domains, subfamily A, member 4, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human MS4A4A protein.
Brand: Abnova H00051338-B02.50uL
Description
This gene encodes a member of the membrane-spanning 4A gene family. Members of this nascent protein family are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues. Alternative splicing of this gene results in several transcript variants; however, not all transcripts have been fully described. [provided by RefSeq
Sequence: MTTMQGMEQAMPGAGPGVPQLGNMAVIHSHLWKGLQEKFLKGEPKVLGVVQILTALMSLSMGITMMCMASNTYGSNPISVYIGYTIWGSVMFIISGSLSIAAGIRTTKGLVRGSLGMNITSSVLAASGILINTFSLAFYSFHHPYCNYYGNSNNCHGTMSILMGLDGMVLLLSVLEFCIAVSLSAFGCKVLCCTPGGVVLILPSHSHMAETASPTPLNEVSpecifications
| membrane-spanning 4-domains, subfamily A, member 4 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human MS4A4A protein. | |
| MS4A4A | |
| 4SPAN1/CD20-L1/CD20L1/HDCME31P/MGC22311/MS4A4/MS4A7 | |
| Mouse | |
| 50 μL | |
| Membrane Receptors | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_024021.2 | |
| MS4A4A | |
| MS4A4A (NP_076926.2, 1 a.a. ∼ 220 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 51338 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.