Learn More
mitochondrial ribosomal protein S27, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human MRPS27 protein.
Brand: Abnova H00023107-B01.50uL
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that may be a functional partner of the death associated protein 3 (DAP3). [provided by RefSeq
Sequence: MPLIWKPGYLDRALQVMEKVAASPEDIKLCREALDVLGAVLKALTSADGASEEQSQNDEDNQGSEKLVEQLDIEETEQSKLPQYLERFKALHSKLQALGKIESEGLLSLTTQLVKEKLSTCEAEDIATYEQNLQQWHLDLVQLIQREQQQREQAKQEYQAQKAAKASASpecifications
| mitochondrial ribosomal protein S27 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human MRPS27 protein. | |
| MRPS27 | |
| FLJ21764/FLJ23348/KIAA0264/MRP-S27/S27mt | |
| Mouse | |
| 50 μL | |
| Stem Cell Biology | |
| Primary | |
| Human | |
| Serum |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| No additive | |
| BC030521 | |
| MRPS27 | |
| MRPS27 (AAH30521, 1 a.a. ∼ 168 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 23107 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.