missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAS-related GPR, member F, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human MRGPRF protein.
Brand: Abnova H00219928-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MAGNCSWEAHPGNRNKMCPGLSEAPELYSRGFLTIEQIAMLPPPAVMNYIFLLLCLCGLVGNGLVLWFFGFSIKRNPFSIYFLHLASADVGYLFSKAVFSILNTGGFLGTFADYIRSVCRVLGLCMFLTGVSLLPAVSAERCASVIFPAWYWRRRPKRLSAVVCALLWVLSLLVTCLHNYFCVFLGRGAPGAACRHMDIFLGILLFLLCCPLMVLPCLALILHVECRARRRQRSAKLNHVILAMVSVFLVSSIYLGIDWFLFWVFQIPAPFPEYVTDLCICINSSAKPIVYFLAGRDKSQRLWEPLRVVFQRALRDGAELGEAGGSTPNTVTMEMQCPPGNASSpecifications
| MAS-related GPR, member F | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human MRGPRF protein. | |
| MRGPRF | |
| GPR140/GPR168/MGC21621/RTA/mrgF | |
| Mouse | |
| 50 μL | |
| Membrane Receptors | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| BC016964 | |
| MRGPRF | |
| MRGPRF (AAH16964, 1 a.a. ∼ 343 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 219928 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction