missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The MHC Class II (I-A/I-E) antibody from Proteintech is a rat monoclonal antibody generated with activated c57bl/6 mouse spleen cells of mouse MHC Class II (I-A/I-E). This antibody recognizes mouse antigen. The MHC Class II (I-A/I-E) antibody has been validated for the following applications: FC analysis.
Specifications
Specifications
| Antigen | MHC Class II (I-A/I-E) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | M5/114.15.2 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Host Species | Rat |
| Immunogen | Activated C57BL/6 mouse spleen cells |
| Purification Method | Affinity purification |
| Quantity | 500 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?