missing translation for 'onlineSavingsMsg'
Learn More

Anti-Mouse MHC Class II (I-A/I-E) (M5/114.15.2), Proteintech

Product Code. 30234131 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μg
500 μg
Unit Size:
100µg
500µg
Product Code. Quantity unitSize
30234131 500 μg 500µg
30236831 100 μg 100µg
2 options
This item is not returnable. View return policy

Product Code. 30234131

Brand: Proteintech 651221IG500UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

The MHC Class II (I-A/I-E) antibody from Proteintech is a rat monoclonal antibody generated with activated c57bl/6 mouse spleen cells of mouse MHC Class II (I-A/I-E). This antibody recognizes mouse antigen. The MHC Class II (I-A/I-E) antibody has been validated for the following applications: FC analysis.

Specifications

Antigen MHC Class II (I-A/I-E)
Applications Flow Cytometry
Classification Monoclonal
Clone M5/114.15.2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Host Species Rat
Immunogen Activated C57BL/6 mouse spleen cells
Purification Method Affinity purification
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14960
Target Species Mouse
Content And Storage Store at 2-8°C. Stable for one year after shipment.
Product Type Flow Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.