missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IGF-I R, Clone: 3B7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-86683JF549
This item is not returnable.
View return policy
Description
c-Myc Monoclonal specifically detects c-Myc in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| IGF-I R | |
| Monoclonal | |
| 1.1mg/mL | |
| Mouse | |
| 0.05 mg | |
| Primary | |
| This reacts specifically with the IGF-1 receptor. This stimulates binding of IGF-1 and -2 to the receptor and stimulates basal and IGF-1 stimulated cell growth. Cross-reacts with Human. Not yet tested in other species. | |
| Purified |
| Immunoprecipitation | |
| 3B7 | |
| Unconjugated | |
| Purified human placental IGF-1 receptors. | |
| RUO | |
| 3480 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction