Learn More
IgG (H+L) Cross-Adsorbed F(ab')2-Rabbit anti-Mouse, Invitrogen™
F(ab')2-Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen PA533287
Description
F(ab')2 fragments were prepared by pepsin digestion of IgG followed by a gel filtration step to remove any remaining intact IgG or Fc fragments. This antibody cross-reacts with rat samples; cross-reactivity with human serum proteins has been minimized by solid phase adsorbtion. This antibody is suitable as a bridge in a PAP/APAAP complex.
Thermo Scientific Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Mouse IgG (H+L) Cross-Adsorbed | |
| Polyclonal | |
| Unconjugated | |
| whole mouse IgG | |
| 1 mg | |
| Secondary | |
| 4°C or -20°C if preferred | |
| IgG |
| ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.