missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Mouse CD69 (H1.2F3), Proteintech
Hamster Monoclonal Antibody
£41.00 - £126.00
Specifications
| Antigen | CD69 |
|---|---|
| Clone | H1.2F3 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235238
|
Proteintech
65105-1-IG-100UG |
100 μg |
£41.70 £41.00 / 100µg Save £0.70 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234124
|
Proteintech
65105-1-IG-500UG |
500 μg |
£129.00 £126.00 / 500µg Save £3.00 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD69 antibody from Proteintech is a armenian hamster monoclonal antibody generated with mouse dendritic epidermal t cell line y245 of mouse CD69. This antibody recognizes mouse antigen. The CD69 antibody has been validated for the following applications: FC analysis.Specifications
| CD69 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 12515 | |
| IgG | |
| Affinity purification | |
| Flow Antibody |
| H1.2F3 | |
| Flow Cytometry | |
| Unconjugated | |
| Armenian Hamster | |
| Mouse | |
| Mouse dendritic epidermal T cell line Y245 | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title