missing translation for 'onlineSavingsMsg'
Learn More

CD43 Rat anti-Mouse, PE, Clone: S7, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 561857

 View more versions of this product

Product Code. 15821409

  • ONLINE EXCLUSIVE PRICE
    £95.25
    £91.34 / Each
    Save £3.91
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The S7 monoclonal antibody specifically binds to the 115 kDa glycosylated form of CD43 (Ly-48, Leukosialin). CD43 is expressed on IL-7-responsive pro-B cells, plasma cells, peritoneal and splenic CD5+ B cells (B-1 cells), granulocytes, monocytes, macrophages, platelets, natural killer cells, thymocytes, peripheral T cytotoxic/suppressor cells, and most T helper cells, but not resting conventional peripheral B cells. CD43 expression has also been detected on pluripotent hematopoietic stem cells and myeloid, lymphoid, and NK-cell progenitors in the bone marrow. Studies of CD43-deficient mice indicate that CD43 participates in the negative regulation of T-cell activation and adhesion.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD43
Monoclonal
0.2mg/mL
Spn ; Sialophorin; Leukosialin; Ly-48; Ly48; Galgp; LEUK
Rat
Affinity Purified
RUO
Mouse
IgG2a κ
Flow Cytometry
S7
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse Plasmacytoma MOPC-315
25 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.