missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Mouse CD1d (1B1), Proteintech
Rat Monoclonal Antibody
£35.00 - £109.00
Specifications
| Antigen | CD1d |
|---|---|
| Clone | 1B1 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235289
|
Proteintech
65242-1-IG-500UG |
500 μg |
£117.00 £109.00 / 500µg Save £8.00 6% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237649
|
Proteintech
65242-1-IG-100UG |
100 μg |
£37.00 £35.00 / 100µg Save £2.00 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD1d antibody from Proteintech is a rat monoclonal antibody generated with mouse cd1d cdna-transfected rma-s mouse t lymphoma of mouse CD1d. This antibody recognizes mouse antigen. The CD1d antibody has been validated for the following applications: FC analysis.Specifications
| CD1d | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 12479 | |
| IgG2b κ | |
| Affinity purification | |
| Flow Antibody |
| 1B1 | |
| Flow Cytometry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Mouse CD1d cDNA-transfected RMA-S mouse T lymphoma | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title