missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Mouse CD11a (M17/4), Proteintech
Rat Monoclonal Antibody
£30.00 - £103.00
Specifications
| Antigen | CD11a |
|---|---|
| Clone | M17/4 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235766
|
Proteintech
65177-1-IG-100UG |
100 μg |
£39.00 £30.00 / 100µg Save £9.00 23% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235834
|
Proteintech
65177-1-IG-500UG |
500 μg |
£119.00 £103.00 / 500µg Save £16.00 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD11a antibody from Proteintech is a rat monoclonal antibody generated with c57bl/6 mouse splenic secondary cytotoxic t cells of mouse CD11a. This antibody recognizes mouse antigen. The CD11a antibody has been validated for the following applications: FC analysis.Specifications
| CD11a | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 16408 | |
| IgG2a κ | |
| Purified by protein-A affinity chromatography | |
| Flow Antibody |
| M17/4 | |
| Flow Cytometry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| C57BL/6 mouse splenic secondary cytotoxic T cells | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title