Learn More
Monocyte/Granulocyte Mouse anti-Pig, PE, Clone: 74-22-15A, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561499
Description
The 74-22-15A (switch variant of 74-22-15) monoclonal antibody, an isotype class-switch variant of mAb 74-22-15, specifically binds to a 230-kDa protein expressed by most pig macrophages, peripheral blood monocytes and granulocytes, and few lymphocytes. mAb 74-22-15A does not crossreact with human or bovine cells. This clone was clustered as anti-SWC3a at the First International Swine CD workshop.
Specifications
| Monocyte/Granulocyte | |
| Monoclonal | |
| 0.2mg/mL | |
| SWC3; CD172a | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Pig | |
| IgG2b κ |
| Flow Cytometry | |
| 74-22-15A | |
| PE | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| dd miniature swine thymocytes | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.