missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Monkey IgG (H+L) Secondary Antibody, HRP
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ PA184631
This item is not returnable.
View return policy
Description
This antibody reacts with Human, African green monkey, Cynomolgus monkey, Baboon, Chimpanzee and Rhesus Monkey IgG. Goat anti Monkey IgG (H/L) antibody recognizes monkey IgG (heavy and light chains).
Anti-Monkey secondary antibodies are affinity-purified antibodies with well-characterized specificity for non-human primate (e.g. monkey) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Monkey IgG (H+L) | |
| Polyclonal | |
| HRP | |
| Goat | |
| Affinity chromatography | |
| RUO | |
| Monkey | |
| Secondary Antibody | |
| IgG |
| ELISA | |
| 1 mg/mL | |
| PBS with 0.2% BSA and 0.05% ProClin 300 | |
| Purified monkey IgG. | |
| 1 mg | |
| Secondary | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction